Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GALNT14 Protein

Catalog No. 9077GT050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-77G-T050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-77G-T050 Supplier R&D Systems™ Supplier No. 9077GT050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to transfer GalNAc from UDP-GalNAc to peptide EA2 from AnaSpec, Inc. The specific activity is >25 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q96FL9
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 79623
Molecular Weight (g/mol) M.W. (Observed): 56-64 kDa, reducing conditions; M.W. (theoretical): 62 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Polypeptide GalNac Transferase 14/GALNT14 protein Thr27-Ser552 with C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 2.4.1.41, FLJ12691, FLJ13977, GalNac-T10, GalNac-T14, GALNT15, Polypeptide GalNAc transferase 14, polypeptide N-acetylgalactosaminyltransferase 14, polypeptide N-acetylgalactosaminyltransferase 15, pp-GaNTase 14, Protein-UDP acetylgalactosaminyltransferase 14, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 14, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T10, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T14, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 14 (GalNAc-T14)
Common Name GALNT14
Gene Symbol GALNT14
Biological Activity Measured by its ability to transfer GalNAc from UDP-GalNAc to peptide EA2 from AnaSpec, Inc. The specific activity is >25 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Thr27-Ser552 with C-terminal 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein GALNT14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.