Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GALT His Protein
SDP

Catalog No. NBP223068 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223068 0.02mg
NB398469 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223068 Supplier Novus Biologicals™ Supplier No. NBP223068
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-379 of Human GALT The Recombinant Human GALT Protein is derived from E. coli. The Recombinant Human GALT Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000146
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.5), 0.2 M NaCl, 10% glycerol
Gene ID (Entrez) 2592
Molecular Weight (g/mol) 45.9kDa
Name GALT Protein
Purification Method SDS-PAGE
Quantity 0.02mg
Source E. Coli
Immunogen GALT, 1-379aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSHMSRSGT DPQQRQQASE ADAAAATFRA NDHQHIRYNP LQDEWVLVSA HRMKRPWQGQ VEPQLLKTVP RHDPLNPLCP GAIRANGEVN PQYDSTFLFD NDFPALQPDA PSPGPSDHPL FQAKSARGVC KVMCFHPWSD VTLPLMSVPE IRAVVDAWAS VTEELGAQYP WVQIFENKGA MMGCSNPHPH CQVWASSFLP DIAQREERSQ QAYKSQHGEP LLMEYSRQEL LRKERLVLTS EHWLVLVPFW ATWPYQTLLL PRRHVRRLPE LTPAERDDLA SIMKKLLTKY DNLFETSFPY SMGWHGAPTG SEAGANWNHW QLHAHYYPPL LRSATVRKFM VGYEMLAQAQ RDLTPEQAAE RLRALPEVHY HLGQKDRETA TIA
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category metabolism
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.