Learn More
Abcam Recombinant Human GAPDH protein, 100UG
Supplier:  Abcam AB77109100UG
Encompass_Preferred
Recombinant Human GAPDH protein is a Human Full Length protein, in the 1 to 335 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: CD194, CMKBR4, CCR4, C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4, CCR-4, K5-5
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2193396
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.