Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Gas6 Protein
SDP

Catalog No. 885GSB050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
885GSB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 885GSB050 Supplier R&D Systems™ Supplier No. 885GSB050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Gas6 Protein is derived from NS0. The Recombinant Human Gas6 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_000811
For Use With (Application) Bioactivity
Gene ID (Entrez) 2621
Molecular Weight (g/mol) Observed M.W.: 70-85kDa, reducing conditions, Theoretical M.W.: 70.5kDakDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Gas6 protein Ala49-­Ala678, with a C-­terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6
Common Name Gas6
Gene Symbol GAS6
Biological Activity Measured by its ability to down regulate the expression of Axl in DU145 human prostate carcinoma cells. Mishra, A. et al. (2012) Mol. Cancer Res. 10:703. The ED50 for this effect is typically 25-150ng/mL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Ala49-­Ala678, with a C-­terminal 6-His tag
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.