Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GCNT1 Protein
SDP

Catalog No. 7248GT020 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7248GT020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 7248GT020 Supplier R&D Systems™ Supplier No. 7248GT020

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to transfer GlcNAc from UDP-GlcNAc to B1-3 galactosyl-N-acetyl galactosamine. The specific activity is >350 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number NP_001481
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Brij-35.
Gene ID (Entrez) 2650
Molecular Weight (g/mol) M.W. (Observed): 50-57 kDa, reducing conditions; M.W. (theoretical): 47 kDa
Name Glucosaminyl (N-acetyl) Transferase 1/GCNT1
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived human Glucosaminyl (N-acetyl) Transferase 1/GCNT1 protein Arg33-His428, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol GCNT1
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.)
Protein Glucosaminyl (N-acetyl) Transferase 1/GCNT1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.