Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GDF-15 (Human Cell-expressed) Protein
SDP

Catalog No. 8146GD010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
8146GD010 10 μg Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
8146GD010CF 10 μg Lyophilized from a 0.2μm filtered solution in HCl.
8146GD025 25 μg Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
8146GD025CF 25 μg Lyophilized from a 0.2μm filtered solution in HCl.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 8146GD010 Supplier R&D Systems™ Supplier No. 8146GD010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its ability to activate SRE-SEAP reporter in HEK293 human embryonic kidney cells transfected with human c-Ret and human GFRAL. The ED50 for this effect is 0.2-2 ng/mL.

The Recombinant Human GDF-15 (Human Cell-expressed) Protein is derived from HEK293.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Gene ID (Entrez) 9518
Molecular Weight (g/mol) 12 kDa (monomer)
Name GDF-15
Quantity 10 μg
Source Human embryonic kidney cell,HEK293-derived human GDF-15 protein Ala197-Ile308
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.