Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GDF-9 Protein

Catalog No. 8266G9010
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
82-66G-9010 Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
82-66G-9010CF Lyophilized from a 0.2μm filtered solution in HCl.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 82-66G-9010 Supplier R&D Systems™ Supplier No. 8266G9010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human GDF-9 Protein is derived from HEK293. The Recombinant Human GDF-9 Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature. Upon receipt, store it immediately

Specifications

Accession Number O60383
Formulation Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Molecular Weight (g/mol) 16 kDa (monomer)
Name GDF-9
Quantity 10 μg
Source Human embryonic kidney cell,HEK293-derived human GDF-9 protein Gly320-Arg454 (Gly391Arg)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.