Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GDNF GMP Protein, CF

Catalog No. RD10008952
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10008952 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10008952 Supplier R&D Systems™ Supplier No. BTGDNFGMP050
Add to Cart
Add to Cart

Recombinant Protein

Measured in a cell proliferation assay using SH-SY5Y human neuroblastoma cells. The ED50 for this effect is 0.750-9.00 ng/mL in the presence of Recombinant Human GFR alpha -1/GDNF R alpha 1 Fc Chimera (Catalog # 714-GR ). The specific activity of recombinant human GDNF is >5.0 x 105 units/mg, which is calibrated against the human GDNF Reference Standard (NIBSC code: 09/266).

Specifications

Accession Number NP_000505.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 2668
Molecular Weight (g/mol) MolecularWeight-observed: 8 kDa, under reducing conditions., MolecularWeight-theroretical: 11.3 kDa
Quantity 50 μg
Source E. coli-derived human GDNF protein Gly113 - Ile211 with & without an N-terminal Met Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidelines.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 12 months when stored at ≤ -20 °C as supplied. Refer to lot specific COA for the Use by Date. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, ≤ -20 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, HGDNF, HSCR3
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in sterile water.
Purity or Quality Grade >97%, by SDS-PAGE with quantitative densitometry by Coomassie™ Blue Staining. The molecular weight by mass spectrometry is 22225 Da ± 5 Da, N-terminal Met species are also present (22356 ± 5 Da)
Protein GDNF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.