Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GDNF Protein

Catalog No. 212GD01M
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
21-2GD-01M Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein
21-2GD-01MCF Lyophilized from a 0.2μm filtered solution in PBS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-2GD-01M Supplier R&D Systems™ Supplier No. 212GD01M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Measured in a cell proliferation assay using SH-SY5Y human neuroblastoma cells. The ED50 for this effect is 2-12 ng/mL in the presence of Recombinant Human GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 714-GR ). Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 714-GR ) at 1 μg/mL can bind Recombinant Human GDNF with an apparent Kd <1 nM.

Order Info

Ships at ambient temperature.

Specifications

Accession Number P39905
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein
Gene ID (Entrez) 2668
Molecular Weight (g/mol) 11.6 kDa (monomer)
Quantity 1 mg
Source Mouse myeloma cell line,NS0-derived human GDNF protein Arg109-Ile211
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method
Biological Activity 2 to 12ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.