Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GIF Protein

Catalog No. 8387TC050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-87T-C050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-87T-C050 Supplier R&D Systems™ Supplier No. 8387TC050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human GIF Protein is derived from HEK293. The Recombinant Human GIF Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P27352
For Use With (Application) Bioactivity
Gene ID (Entrez) 2694
Molecular Weight (g/mol) Observed M.W.: 44-60kDa, reducing conditions, Theoretical M.W.: 44kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human GIF protein Ser19-Tyr417 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias gastric intrinsic factor (vitamin B synthesis), IF, IFIntrinsic factor, IFMH, IFMHgastric intrinsic factor, INF, TCN3
Common Name GIF
Gene Symbol GIF
Biological Activity Measured by its ability to bind Vitamin B12. The specific binding is =30μg Vitamin B12/mg Recombinant Human GIF.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Ser19-Tyr417 with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.