Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GIPR Fc Chimera Protein

Catalog No. 11088GI050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11088GI050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11088GI050 Supplier R&D Systems™ Supplier No. 11088GI050
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA. When Biotinylated Human GIP Peptide is captured on a Streptavidin Coated Plate (Catalog # CP004 ), it binds to Recombinant Human GIPR Fc Chimera. The ED50 for this binding is 20.0-100 ng/mL.

Specifications

Accession Number P48546.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 2696
Molecular Weight (g/mol) MolecularWeight-observed: 45-59 kDa, under reducing conditions., MolecularWeight-theroretical: 40 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human GIPR protein Human GIPR (Gly26-Gln138) (N-terminus) Accession # P48546.1 IEGRMD Human IgG1 Fc (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein GIPR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.