Learn More
R&D Systems™ Recombinant Human GITR Ligand Alexa Fluor™ 647 Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Accession Number | Q9UNG2.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 8995 |
| Molecular Weight (g/mol) | M.W.-observed: 20-27 kDa, under reducing conditions., M.W.-theroretical: 19.6 kDa (monomer) |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human GITR Ligand/TNFSF18 protein HA (YPYDVPDYA) GCN4-IZ Human GITR Ligand(Glu52-Ser177)(C-terminus) Accession # Q9UNG2.3 (N-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.