Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GLTPD1 His Protein
SDP

Catalog No. NBP25842002 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25842002 0.02 mg
NBP25184201 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25842002 Supplier R&D Systems™ Supplier No. NBP2518420.02MG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-214 of Human GLTPD1 The Recombinant Human GLTPD1 Protein is derived from E. coli. The Recombinant Human GLTPD1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH7.4) containing 50% glycerol, 1mM DTT
Gene ID (Entrez) 80772
Molecular Weight (g/mol) TMW: 26.8kDa
Quantity 0.02 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias Ceramide-1-Phosphate Transfer Protein, CPTP, Glycolipid Transfer Protein Domain Containing 1, Glycolipid Transfer Protein Domain-Containing Protein 1
Common Name GLTPD1
Conjugate Unconjugated
Recombinant Recombinant
Product Line Novus
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.