Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Glutathione Reductase Protein

Catalog No. 8866GR100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8866GR100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8866GR100 Supplier R&D Systems™ Supplier No. 8866GR100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Glutathione Reductase Protein is derived from E. coli. The Recombinant Human Glutathione Reductase Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P00390
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 2936
Molecular Weight (g/mol) Observed M.W.: 53kDa, reducing conditions, Theoretical M.W.: 53kDa
Name SPR
Quantity 100 μg
Source E. coli-derived human Glutathione Reductase protein Met44-Arg522,with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias Glutathione Reductase
Common Name Glutathione Reductase
Gene Symbol GSR
Biological Activity Measured by its ability to reduce oxidized glutathione to glutathione in an NADPH-dependent manner. The specific activity is >15,000 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Met44-Arg522, with an N-terminal Met and 6-His tag Accession # P00390
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5μg per lane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.