Learn More
R&D Systems™ Recombinant Human Glypican 4 Fc Chimera Protein
Description
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| Accession Number | NP_001019520 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and NaCl. |
| Gene ID (Entrez) | 2239 |
| Molecular Weight (g/mol) | 84 kDa, reducing conditions |
| Name | glypican 4, glypican proteoglycan 4, glypican-4, GPC4, K-glypican, K-glypicandJ900E8.1 (glypican 4) |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Glypican 4 protein, (N-terminus) Human Glypican 4 (Ala23-Ser529) Accession &Num; O75487 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.