Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GM-CSF Protein
GSA_VA

Catalog No. 215GM050CF
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
215GM050CF 50 μg Lyophilized from a 0.2μm filtered solution in PBS.
215GM010 10 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
215GM050 50 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
215GM010CF 10 μg Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 215GM050CF Supplier R&D Systems™ Supplier No. 215GM050/CF
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 6-30 pg/mL.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accession Number P04141.1
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 1437
Molecular Weight (g/mol) 14 kDa
Name GM-CSF
Quantity 50 μg
Source E. coli-derived human GM-CSF protein Ala18-Glu144
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.