Learn More
Description
Specifications
Specifications
| Concentration | 0.25mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 0.2M NaCl, 40% glycerol, 2mM DTT, 0.1mM PMSF |
| Gene ID (Entrez) | 51764 |
| Molecular Weight (g/mol) | TMW: 10kDa |
| Quantity | 0.02 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | G gamma subunit, clone:h2-35, G(gamma)13, guanine nucleotide binding protein (G protein), gamma 13, guanine nucleotide binding protein 13, gamma, guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-13, h2-35 |
| Common Name | GNG13 |
| Conjugate | Unconjugated |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
