Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human gp130 Protein

Catalog No. 228GP050CF
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and Trehalose.
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
228GP050CF 50 μg Lyophilized from a 0.2μm filtered solution in PBS and Trehalose.
228GP010CF 10 μg Lyophilized from a 0.2μm filtered solution in PBS and Trehalose.
228GP010 10 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
228GP050 50 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 228GP050CF Supplier R&D Systems™ Supplier No. 228GP050/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human gp130 Protein is derived from Sf 21 (baculovirus). The Recombinant Human gp130 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS and Trehalose.
Gene ID (Entrez) 3572
Molecular Weight (g/mol) 68 kDa
Name gp130
Quantity 50 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human gp130 protein Leu24-Glu619 (Glu619Asp)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.