Learn More
Specifications
Specifications
| Concentration | 0.25 mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | PBS (pH 7.4), 10% glycerol |
| Gene ID (Entrez) | 2812 |
| Molecular Weight (g/mol) | 14 kDa |
| Quantity | 50 μg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | < 1 EU per 1ug of protein (determined by LAL method) |
| Gene Alias | antigen CD42b-beta, BDPLT1, BS, glycoprotein Ib (platelet), beta polypeptide, glycoprotein Ib platelet beta subunit, GPIBB, GPIbbeta, GPIb-beta, nuclear localization signal deleted in velocardiofacial syndrome, platelet glycoprotein Ib beta chain, platelet membrane glycoprotein Ib beta, truncated platelet membrane glycoprotein Ib beta |
| Gene Symbol | GP1BB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
