Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GPR87 Protein
SDP

Catalog No. NB398879 Shop All R&D Systems Products
Change view
Click to view available options
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NB398879 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398879 Supplier Novus Biologicals™ Supplier No. H00053836G01
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) Immunoaffinity Purification
Formulation 25 mM Tris-HCl pH8.0 in 2% glycerol.
Gene ID (Entrez) 53836
Molecular Weight (g/mol) 39.4 kDa
Quantity 2 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias G protein-coupled receptor 87, G protein-coupled receptor 95, GPR95, G-protein coupled receptor 87, G-protein coupled receptor 95, KPG_002, MGC131898, orphan GPCR 87
Gene Symbol GPR87
Research Category GPCR
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein GPR87
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.