Learn More
Sartorius Recombinant Human Granulocyte-macrophage colony-stimulating factor
Shop All Sartorius ProductsDescription
- Research Use Only
Specifications
Specifications
| For Use With (Application) | Organoid Expansion |
| Quantity | 0.1 mg |
| Product Type | Recombinant Proteins |
| Conjugate | Unconjugated |
| Species | Human |
| Recombinant | Recombinant |
| Form | Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.