Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Granzyme A Protein

Catalog No. 2905SE010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2905SE010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2905SE010 Supplier R&D Systems™ Supplier No. 2905SE010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Granzyme A Protein is derived from NS0. The Recombinant Human Granzyme A Protein has been validated for the following applications: Enzyme Activity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in MES, NaCl and CaCl 2 with Trehalose.
Gene ID (Entrez) 3001
Molecular Weight (g/mol) 28 kDa
Name Granzyme A
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived human Granzyme A protein Cys26-Val262,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Pro form
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 25 mM HEPES, 150 mM NaCl and 10 mM CaCl 2 pH 7.5.
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.