Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Growth Hormone R Protein
SDP

Catalog No. NB002064 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002064 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002064 Supplier Novus Biologicals™ Supplier No. NBP19990650UG

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant bioactive protein containing 237 amino acids for Human Growth Hormone R The Recombinant Human Growth Hormone R Protein is derived from E. coli. The Recombinant Human Growth Hormone R Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Lyophilized from 0.0045 mM NaHCO3.
Gene ID (Entrez) 2690
Molecular Weight (g/mol) 30.3 kDa
Quantity 50 μg
Source E. coli
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias GH receptor, GHBP, growth hormone binding protein, growth hormone receptor, serum binding protein, Somatotropin receptor
Gene Symbol GHR
Reconstitution Reconstitute in sterile dH2O to no less than 100 μg/mL. This solution can then be diluted into other aqueous buffers.
Buffer Lyophilized from 0.0045 mM NaHCO3.
Endotoxin Level <0.1 ng/μg
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.