Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GRP78/HSPA5 His Protein
SDP

Catalog No. NB171979 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171979 50 μg
NB171977 100 μg
NB171978 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB171979 Supplier Novus Biologicals™ Supplier No. NBP31831950UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

The protein has ATPase activity at the time of manufacture of 2.3uM phosphate liberated/hr/ug protein in a 200ul reaction at 37C (pH 8) in the presence of 20ul of 1mM ATP using a Malachite Green assay.

Specifications

For Use With (Application) Bioactivity, ELISA, Functional Assay, SDS-PAGE, Western Blot
Formulation 20mM Tris/HCl pH7.5, 50mM KCl, 5mM MgCl2, 1mM DTT
Gene ID (Entrez) 3309
Molecular Weight (g/mol) M.W. Theoretical: 78 kDa
Quantity 50 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2
Gene Symbol HSPA5
Research Category Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR)
Purity or Quality Grade >85%
Protein GRP78/HSPA5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.