Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GRP78/HSPA5 His Protein
SDP

Catalog No. NBP223104 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223104 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223104 Supplier Novus Biologicals™ Supplier No. NBP223104
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 19-654 of Human GRP78/HSPA5 The Recombinant Human GRP78/HSPA5 Protein is derived from E. coli. The Recombinant Human GRP78/HSPA5 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_005338
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 3309
Molecular Weight (g/mol) 72.9kDa
Name GRP78/HSPA5 Protein
Purification Method SDS-PAGE
Quantity 0.1mg
Source E. Coli
Immunogen HSPA5, 19-654aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSEEEDKKE DVGTVVGIDL GTTYSCVGVF KNGRVEIIAN DQGNRITPSY VAFTPEGERL IGDAAKNQLT SNPENTVFDA KRLIGRTWND PSVQQDIKFL PFKVVEKKTK PYIQVDIGGG QTKTFAPEEI SAMVLTKMKE TAEAYLGKKV THAVVTVPAY FNDAQRQATK DAGTIAGLNV MRIINEPTAA AIAYGLDKRE GEKNILVFDL GGGTFDVSLL TIDNGVFEVV ATNGDTHLGG EDFDQRVMEH FIKLYKKKTG KDVRKDNRAV QKLRREVEKA KRALSSQHQA RIEIESFYEG EDFSETLTRA KFEELNMDLF RSTMKPVQKV LEDSDLKKSD IDEIVLVGGS TRIPKIQQLV KEFFNGKEPS RGINPDEAVA YGAAVQAGVL SGDQDTGDLV LLDVCPLTLG IETVGGVMTK LIPRNTVVPT KKSQIFSTAS DNQPTVTIKV YEGERPLTKD NHLLGTFDLT GIPPAPRGVP QIEVTFEIDV NGILRVTAED KGTGNKNKIT ITNDQNRLTP EEIERMVNDA EKFAEEDKKL KERIDTRNEL ESYAYSLKNQ IGDKEKLGGK LSSEDKETME KAVEEKIEWL ESHQDADIED FKAKKKELEE IVQPIISKLY GSAGPPPTGE EDTAEKDEL
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.