Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human GSTA1 Protein
SDP

Catalog No. NBP130307 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130307 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130307 Supplier R&D Systems™ Supplier No. NBP130307

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-222 of Human GSTA1 The Recombinant Human GSTA1 Protein is derived from E. coli. The Recombinant Human GSTA1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number P08263
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 1mM DTT, 10% glycerol
Gene ID (Entrez) 2938
Molecular Weight (g/mol) 25.6kDa
Name GSTA1 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen GSTA1, 1-222aa, Human, E.coli (NP_665683).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Product Line Novus
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.