Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Guanine deaminase Protein
SDP

Catalog No. NB2496925UG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
5 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB2496925UG 5 μg
NB24969220 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB2496925UG Supplier Novus Biologicals™ Supplier No. NBP2496925UG
Only null left
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-454 of Human Guanine deaminase The Recombinant Human Guanine deaminase Protein is derived from E. coli. The Recombinant Human Guanine deaminase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 1mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol, 1mM DTT
Gene ID (Entrez) 9615
Molecular Weight (g/mol) 51 kDa
Name Recombinant Human Guanine deaminase Protein
Purification Method >90%, by SDS-PAGE
Quantity 5 μg
Source E.coli
Storage Requirements −80°C in dark, avoid freeze-thaw cycles
Regulatory Status RUO
Gene Alias CYPIN, EC 3.5.4.3, GAH, GUANASE, Guanine aminase, Guanine aminohydrolase, guanine deaminase, KIAA1258, MGC9982, NEDASIN, p51-nedasin
Gene Symbol GDA
Product Type Recombinant protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.