Learn More
R&D Systems™ Recombinant Human HA-Ubiquitin Protein
Description
Specifications
Specifications
| Accession Number | P0CG47.1 |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES. |
| Gene ID (Entrez) | 7314 |
| Molecular Weight (g/mol) | 9.8 kDa |
| Name | HA Ubiquitin |
| Quantity | 1mg |
| Source | E. coli-derived human Ubiquitin protein Met1 - Gly76 with N-terminal HA (YPYDVPDYA) tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.