Learn More
R&D Systems™ Recombinant Human HABP1/C1QBP Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in MOPS and NaCl. |
| Gene ID (Entrez) | 708 |
| Molecular Weight (g/mol) | 24.7 kDa |
| Name | HABP1/C1QBP |
| Quantity | 50 μg |
| Source | E. coli-derived human HABP1/C1QBP protein His75-Gln282,with an N-term 2-Met tag and a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute at 100 μg/mL in sterile deionized water. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.