Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HADHB His Protein
SDP

Catalog No. NBP25194805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25194805 0.5 mg
NBP25194801 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25194805 Supplier Novus Biologicals™ Supplier No. NBP2519480.5MG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 34-474 of Human HADHB The Recombinant Human HADHB Protein is derived from E. coli. The Recombinant Human HADHB Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM phosphate (pH8.0) containing 10% glycerol
Gene ID (Entrez) 3032
Molecular Weight (g/mol) TMW: 49.9kDa
Quantity 0.5 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias 2-enoyl-Coenzyme A (CoA) hydratase, beta subunit, 3-ketoacyl-Coenzyme A (CoA) thiolase of mitochondrial trifunctional protein, acetyl-CoA acyltransferase, beta subunit, beta-ketothiolase, EC 2.3.1, EC 2.3.1.16, ECHB, hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase(trifunctional protein), beta subunit, hydroxyacyl-Coenzyme A (CoA) dehydrogenase, beta subunit, hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme Athiolase/enoyl-Coenzyme A hydratase (trifunctional protein), beta subunit, MGC87480, mitochondrial trifunctional enzyme, beta subunit, mitochondrial trifunctional protein, beta subunit, MTPB, TP-beta, trifunctional enzyme subunit beta, mitochondrial
Common Name HADHB
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.