Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HAI-2/SPINT2 His Protein
SDP

Catalog No. p-7105013 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25214005 0.05 mg
NBP25211425 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25214005 Supplier Novus Biologicals™ Supplier No. NBP2521140.05MG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 28-197 of Human HAI-2/SPINT2 The Recombinant Human HAI-2/SPINT2 Protein is derived from Baculovirus. The Recombinant Human HAI-2/SPINT2 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.2mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH 7.4), 10% glycerol
Gene ID (Entrez) 10653
Molecular Weight (g/mol) TMW: 20kDa
Quantity 0.05 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2
Common Name HAI-2/SPINT2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.