Learn More
R&D Systems™ Recombinant Human HB-EGF (E. coli-expressed) Protein, CF
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in NaPO4 and NaCl. |
| Gene ID (Entrez) | 1839 |
| Molecular Weight (g/mol) | 8 to 12 kDa, under reducing conditions |
| Quantity | 100 μg |
| Source | E. coli-derived human HB-EGF protein Asp63-Leu148 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), Dtr, DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), Dts, DTSF, HBEGF, Hegfl, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.