Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HB-EGF Protein

Catalog No. 259HE050CF
Change view
Click to view available options
Quantity:
250 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
259HE050CF 50 μg Lyophilized from a 0.2μm filtered solution in PBS.
259HE050 50 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
259HE250 250 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
259HE250CF 250 μg Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 259HE050CF Supplier R&D Systems™ Supplier No. 259HE050/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human HB-EGF Protein is derived from Sf 21 (baculovirus). The Recombinant Human HB-EGF Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 1839
Molecular Weight (g/mol) Theoretical Mole. Wt.: 9.7 kDa
Name HB-EGF
Quantity 50 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human HB-EGF protein Asp63-Leu148
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE with silver staining.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.