Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HECT domain-containing 2 Protein, CF
SDP

Catalog No. E3302025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3302025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E3302025 Supplier R&D Systems™ Supplier No. E3302025

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Typical enzyme concentration to support conjugation in vitro is 100-500 nM depending on experimental conditions.

Specifications

Host Species Human
Purity >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Storage Buffer X mg/mL (X μM) in 50 mM HEPES, pH 7.0, 100 mM NaCl, 5% (v/v) Glycerol, 5 mM DTT
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -70°C as supplied. 6 months, -70°C under sterile conditions after opening.
Accession Number Q5U5R9
Gene ID (Entrez) 143279
Gene Alias EC 6.3.2.-, EC2.3.2.26, FLJ16050, FLJ37306, HECT domain containing 2, HECT domain-containing protein 2, probable E3 ubiquitin-protein ligase HECTD2
Conjugate Unconjugated
Source E. coli-derived human HECTD2 protein
Quantity 25 μg
Molecular Weight (g/mol) 88 kDa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.