Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Heparan Sulfate-6-O-Sulfotransferase-3
SDP

Catalog No. 8568ST020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-68S-T020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 85-68S-T020 Supplier R&D Systems™ Supplier No. 8568ST020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to transfer sulfate from PAPS to heparan sulfate. The specific activity is >65 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q8IZP7
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 266722
Molecular Weight (g/mol) M.W. (Observed): 60-95 kDa, reducing conditions; M.W. (theoretical): 52 kDa
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3 protein Pro28-Trp471, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias DKFZp761K2315, heparan sulfate 6-O-sulfotransferase 3
Common Name Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3
Gene Symbol HS6ST3
Biological Activity Measured by its ability to transfer sulfate from PAPS to heparan sulfate. The specific activity is >65 pmol/min/μg, as measured under the described conditions.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Pro28-Trp471, with an N-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.)
Protein Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.