Learn More
R&D Systems™ Recombinant Human Hepassocin/FGL1 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q08830.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 2267 |
| Molecular Weight (g/mol) | M.W.-observed: 60-65 kDa, under reducing conditions, M.W.-theroretical: 61 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human Hepassocin/FGL1 protein MD Human IgG1 (Pro100-Lys330) IEGR Human Hepassocin/FGF1 (Leu23-Ile312) (c-terminus) Accession # Q08830.3 N-terminus C-terminμs |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | FGL1, fibrinogen-like 1, fibrinogen-like protein 1, Fibrinogen-like-protein 1, FREP1, Hepassocin, hepatocellular carcinoma-related sequence, Hepatocyte-derived fibrinogen-related protein 1, HFREP-1, HFREP1MGC12455, HP-041, LFIRE1, LFIRE-1, Liver fibrinogen-related protein 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.