Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HGF (NS0-expressed) Protein

Catalog No. 294HGN005CF
Change view
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
294HGN005CF 5 μg Lyophilized from a 0.2μm filtered solution in PBS.
294HGN005 5 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
294HGN025 25 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
294HGN025CF 25 μg Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 294HGN005CF Supplier R&D Systems™ Supplier No. 294HGN005/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

  • Source: Mouse myeloma cell line, NS0-derived, Gln32-Ser728
  • Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight: alpha chain: 53.7 kDabeta chain: 26 kDa
  • Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
  • Entrez Gene ID: 3082
  • Regulatory Status: RUO

The Recombinant Human HGF (NS0-expressed) Protein is derived from NS0. The Recombinant Human HGF (NS0-expressed) Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3082
Molecular Weight (g/mol) alpha chain: 53.7 kDabeta chain: 26 kDa
Name HGF
Quantity 5 μg
Source Mouse myeloma cell line, NS0-derived human HGF protein Gln32-Ser728 (Asp384Asn, Asp416Asn, Leu622Ser, His645Arg, Val678Ile)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked heterodimer
Gene Alias deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.