Learn More
R&D Systems™ Recombinant Human HGF (NS0-expressed) Protein
Description
- Source: Mouse myeloma cell line, NS0-derived, Gln32-Ser728
- Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
- Molecular Weight: alpha chain: 53.7 kDabeta chain: 26 kDa
- Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
- Entrez Gene ID: 3082
- Regulatory Status: RUO
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3082 |
| Molecular Weight (g/mol) | alpha chain: 53.7 kDabeta chain: 26 kDa |
| Name | HGF |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line, NS0-derived human HGF protein Gln32-Ser728 (Asp384Asn, Asp416Asn, Leu622Ser, His645Arg, Val678Ile) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Gene Alias | deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.