Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HGF Protein, CF
SDP

Catalog No. 294HG100CF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-4HG-100CF 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-4HG-100CF Supplier R&D Systems™ Supplier No. 294HG100/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its ability to induce IL-11 secretion by Saos-2 human osteosarcoma cells. The specific activity of Recombinant Human HGF is approximately 1.5 x 103 IU/μg, which is calibrated against recombinant human HGF WHO International Standard (NIBSC code: 96/564).

  • The ED50 for this effect is 0.1 to 0.5 ng/mL.
  • The specific activity of Recombinant Human HGF is approximately 1.5 x 103 IU/μg, which is calibrated against recombinant human HGF WHO International Standard (NIBSC code: 96/564).

Specifications

Accession Number P14210.2
For Use With (Application) Bioactivity
Gene ID (Entrez) 3082
Molecular Weight (g/mol) 53.7 kDa ( alpha chain), 26 kDa ( beta chain)
Name deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A
Quantity 100 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human HGF protein Gln32-Ser728 (Asp384Asn, Asp416Asn, Leu622Ser, His645Arg, Val678Ile)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol HGF
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in NaH2PO4 and NaCl.
Endotoxin Level <1.0 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.