Learn More
R&D Systems™ Recombinant Human HGFR/c-MET Fc Chimera His-tag Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 4233 |
| Molecular Weight (g/mol) | 32.5 kDa (alpha chain), 96.7 kDa (beta chain) |
| Name | HGFR/c-MET |
| Quantity | 100 μg |
| Source | Mouse myeloma cell line,NS0-derived human HGF R/c-MET protein, (N-terminus) HGF R alpha (Glu25-Arg307) Accession &Num; P08581 HGF R beta (Ser308-Thr932) Accession &Num; P08581 HIEGRMD Human IgG1 (Pro100-Lys330) 6 His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Tetramer containing two disulfide-linked proteolytically cleaved alpha and beta subunits |
| Gene Alias | AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.