Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HGFR/c-MET Fc Chimera Protein
SDP

Catalog No. 8614MT100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-14M-T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 86-14M-T100 Supplier R&D Systems™ Supplier No. 8614MT100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human HGF R/c-MET Fc Chimera Protein is derived from HEK293. The Recombinant Human HGF R/c-MET Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 4233
Molecular Weight (g/mol) Observed M.W.: 34-48kDa and 103-126kDa, reducing conditions, Theoretical M.W.: 33kDa & 96kDa
Name HGFR/c-MET
Quantity 100 μg
Source Human embryonic kidney cell,HEK293-derived human HGF R/c-MET protein, (N-terminus) Human HGF R alpha (Glu25-Arg307) Accession &Num; P08581 Human HGF R beta (Ser308-Thr932) Accession &Num; P08581 HIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met
Common Name HGF R/c-MET
Gene Symbol MET
Biological Activity Measured by its ability to bind immobilized recombinant human HGF in a functional ELISA with an estimated Kd <0.15 nM.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human HGF R alpha (Glu25-Arg307) Accession # P08581, N-terminus; Human HGF R beta (Ser308-Thr932) Accession # P08581; HIEGRMD; Human IgG1 (Pro100-Lys333), C-terminus
Purity or Quality Grade >85%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.