Learn More
R&D Systems™ Recombinant Human Hip Protein
Description
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| Accession Number | Q96QV1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, DTT and Glycerol |
| Gene ID (Entrez) | 64399 |
| Molecular Weight (g/mol) | 77 to 89 kDa, reducing conditions |
| Name | FLJ90230, hedgehog interacting protein, hedgehog-interacting protein, HHIP, HIPFLJ20992 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Hip protein, (N-terminus) Human Hip (Phe18-Asp670) Accession &Num; Q96QV1 HP GGGSGGGSGGGS HHHHHH (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.