Learn More
R&D Systems™ Recombinant Human His10-cIAP-2/HIAP-1 Protein, CF
Description
Specifications
Specifications
| Accession Number | Q13489 |
| Gene ID (Entrez) | 330 |
| Molecular Weight (g/mol) | 71 kDa |
| Name | AIP1, API2, BIRC3, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, HIAP-1, MALT2, MIHC, RNF49 |
| Quantity | 50 μg |
| Source | E. coli-derived human cIAP-2/HIAP-1 protein Contains an N-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.