Learn More
R&D Systems™ Recombinant Human His10-USP24 Protein, CF
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | X mg/mL (X μM) is 50 mM HEPES, pH 8.0, 100 mM NaCl, 10% (v/v) Glycerol, 5 mM DTT, 1 mM EDTA |
| Gene ID (Entrez) | 23358 |
| Molecular Weight (g/mol) | M.W. (theoretical): 296 kDa |
| Name | Deubiquitinating enzyme 24, EC 3.4.19.12, FLJ31309, KIAA1057ubiquitin carboxyl-terminal hydrolase 24, ubiquitin specific peptidase 24, ubiquitin specific protease 24, ubiquitin thioesterase 24, Ubiquitin thiolesterase 24, ubiquitin-specific processing protease 24, Ubiquitin-specific-processing protease 24 |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP24 protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.