Learn More
Description
Specifications
Specifications
| Accession Number | Q05086.4 |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl and TCEP. |
| Gene ID (Entrez) | 7337 |
| Molecular Weight (g/mol) | M.W. (theoretical): 102 kDa |
| Name | E6AP/UBE3A |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human E6AP/UBE3A protein Met1 - Leu875 with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Gene Symbol | UBE3A |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
