Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-Fluorescein-Tandem UBA (TUBE1), CF

Catalog No. UBE212100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UBE212100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. UBE212100 Supplier R&D Systems™ Supplier No. UBE212100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human His6-Fluorescein-Tandem UBA (TUBE1) is derived from E. coli. The Recombinant Human His6-Fluorescein-Tandem UBA (TUBE1) has been validated for the following applications: Bioactivity.

Specifications

Accession Number P54725
For Use With (Application) Bioactivity
Gene ID (Entrez) 5886
Molecular Weight (g/mol) 29 kDa
Quantity 100 μg
Source E. coli-derived
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
Regulatory Status RUO
Gene Alias hHR23, hHR23A, HR23A, Rad23A
Specific Reactivity HR23A/Rad23A TUBE1
Conjugate Fluorescein
Species Human
Recombinant Recombinant
Buffer X mg/mL (X μm) in 50 mM HEPES pH 7.5, 200 mM NaCl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.