Learn More
R&D Systems™ Recombinant Human His6-HSP40/DNAJB1 Protein
Description
Specifications
Specifications
| Gene ID (Entrez) | 3337 |
| Name | His6 HSP40/DNAJB1 |
| Quantity | 50 μg |
| Source | E. coli-derived human HSP40/DNAJB1 protein Contains a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 6 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.