Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-Livin alpha Protein, CF

Catalog No. E3295050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3295050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E3295050 Supplier R&D Systems™ Supplier No. E3295050
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Recombinant Human Livin alpha is Ubiquitin (E3) ligase that functions downstream of a Ubiquitin-activating (E1) enzyme and a Ubiquitin-conjugating (E2) enzyme to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application.

Specifications

Accession Number Q96CA5-1
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 7.5, 150 mM NaCl, 1 mM DTT
Gene ID (Entrez) 79444
Molecular Weight (g/mol) M.W. (theoretical): 34 kDa
Name baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, BIRC7, KIAP, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, ML-IAP, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7
Quantity 50 μg
Source E. coli-derived human Livin protein Contains a C-terminal 6-His tag.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Conjugate Unconjugated
Species Human
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein Livin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.