Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-NEDD4 Protein
SDP

Catalog No. E3290050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3290050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E3290050 Supplier R&D Systems™ Supplier No. E3290050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Recombinant Human NEDD 4 is a Ubiquitin Ligase (E3) that functions downstream of a Ubiquitin-activating (E1) enzyme and a Ubiquitin-conjugating (E2) enzyme to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application.

Specifications

Accession Number P46934
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 8.0, 100 mM NaCl, 5 mM DTT, 1 mM EDTA
Gene ID (Entrez) 4734
Molecular Weight (g/mol) M.W. (theoretical): 88 kDa
Name Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1
Quantity 50 μg
Source E. coli-derived human NEDD4 protein Contains a N-terminal 6-His tag, Leu570 - Asp1319
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Conjugate Unconjugated
Species Human
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein NEDD4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.