Learn More
R&D Systems™ Recombinant Human His6-OTUD6A Protein, CF
Description
Specifications
Specifications
| Accession Number | Q8L7S5 |
| For Use With (Application) | Bioactivity |
| Formulation | X mg/mL (X μM) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 1 mM TCEP |
| Gene ID (Entrez) | 139562 |
| Molecular Weight (g/mol) | M.W. (theoretical): 33 kDa |
| Purification Method | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
| Quantity | 50 μg |
| Source | Trichoplusia ni, T. ni (baculovirus)-derived human OTUD6A protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.